Learn More
MSC Milwaukee Tool Plate Compactor Water Tank Kit, 3.6 gal, MXF220 Compatible
Catalog No. 502838945
Encompass
The Plate Compactor Water Tank Kit is a removable equipment-mounted water tank designed for use with the MX FUEL 20" Plate Compactor. This kit includes a 3.6-gallon water tank and bar with an adjustable flow-rate dial, plus a mounting strap for easy on-the-job tank removal and installation. The tank slides onto the front frame of the plate compactor, secured by the mounting strap, making it ideal for asphalt and various general compaction applications. The adjustable flow rate dial ensures precise control and optimization of water flow through the bar.
  • 3.6 gal (13.6 L) tank capacity
  • Max flow rate of 1 GPM (3.8 LPM)
  • Adjustable flow rate dial for precise control
  • Mounting strap for simple on-the-job tank removal and installation
  • Designed for use with MX FUEL 20" Plate Compactor
  • Constructed from plastic
Catalog No. 50-283-8945 Supplier MSC Supplier No. 60548419
The Plate Compactor Water Tank Kit is a removable equipment-mounted water tank designed for use with the MX FUEL 20" Plate Compactor. This kit includes a 3.6-gallon water tank and bar with an adjustable flow-rate dial, plus a mounting strap for easy on-the-job tank removal and installation. The tank slides onto the front frame of the plate compactor, secured by the mounting strap, making it ideal for asphalt and various general compaction applications. The adjustable flow rate dial ensures precise control and optimization of water flow through the bar.
  • 3.6 gal (13.6 L) tank capacity
  • Max flow rate of 1 GPM (3.8 LPM)
  • Adjustable flow rate dial for precise control
  • Mounting strap for simple on-the-job tank removal and installation
  • Designed for use with MX FUEL 20" Plate Compactor
  • Constructed from plastic