Learn More
MOLTOX Molecular Toxicology Inc Plate Count Agar, 25 mL/plate, 20 plates/sleeve, Store at Room Temperature
SDP
Catalog No. 50948667
Encompass_Preferred
Plate Count Agar, or Standard Methods Agar, is used for the enumeration of bacteria in a variety of samples including dairy products, food, water and other materials. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-667 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S46.141
Plate Count Agar, or Standard Methods Agar, is used for the enumeration of bacteria in a variety of samples including dairy products, food, water and other materials. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.