Learn More
Alkali Scientific Plate Count Agar (Standard Methods), 5 Kilograms
SDP
Catalog No. 5000661542
Encompass_Preferred
Plate Count Agar (Standard Methods), 5kg, is the recognized medium for determining the total viable count of microorganisms in food, water, and dairy products. This medium supports the growth of a wide range of bacteria, helping researchers and quality control labs assess the microbial quality of products. The 5kg pack size is ideal for laboratories involved in large-scale testing or environmental monitoring. The formulation provides nutrients to support the growth of both aerobic and facultative anaerobic organisms, allowing it to serve as a comprehensive medium for bacterial enumeration in various microbiological analyses.
Catalog No. 50-006-61542 Supplier Alkali Scientific Supplier No. PLA001CP
Plate Count Agar (Standard Methods), 5kg, is the recognized medium for determining the total viable count of microorganisms in food, water, and dairy products. This medium supports the growth of a wide range of bacteria, helping researchers and quality control labs assess the microbial quality of products. The 5kg pack size is ideal for laboratories involved in large-scale testing or environmental monitoring. The formulation provides nutrients to support the growth of both aerobic and facultative anaerobic organisms, allowing it to serve as a comprehensive medium for bacterial enumeration in various microbiological analyses.