Learn More
HIMEDIA Plate Count Agar (Standard Methods Agar), M091, Microbiology Culture Media, Dehydrated Culture Media, Agar, Aerobic Plate Count, Food Water and Wastewater, Powder, 500 g
Catalog No. NC4013664
Encompass_Preferred
Plate Count Agar, also known as Standard Methods Agar, is the reference medium for determining aerobic plate counts of micro-organisms in food, water and wastewater. Its simple balanced formulation of tryptone, yeast extract, dextrose and agar, final pH 7.0, supports the growth of a broad range of heterotrophic bacteria for enumeration by the pour-plate or spread-plate method. Trusted in routine quality control for total viable count testing across food, dairy and environmental laboratories. Dehydrated powder, store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013664 Supplier HIMEDIA Supplier No. M091500G
Plate Count Agar, also known as Standard Methods Agar, is the reference medium for determining aerobic plate counts of micro-organisms in food, water and wastewater. Its simple balanced formulation of tryptone, yeast extract, dextrose and agar, final pH 7.0, supports the growth of a broad range of heterotrophic bacteria for enumeration by the pour-plate or spread-plate method. Trusted in routine quality control for total viable count testing across food, dairy and environmental laboratories. Dehydrated powder, store in a cool dry place. Packaged as 500 g.