Learn More
Sigma Aldrich Fine Chemicals Biosciences X-Pierce Film, 100/pk
Catalog No. NC1109594
Encompass
Also useful for protecting contents of plate from contamination during multiple manual reagent additions over time. X-Pierce films are 89 um thick vinyl with a 18 um thick adhesive layer, designed for temporary protection from contamination and evaporation for samples in 96 well plates.
Catalog No. NC1109594 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z722502100EA
Also useful for protecting contents of plate from contamination during multiple manual reagent additions over time. X-Pierce films are 89 um thick vinyl with a 18 um thick adhesive layer, designed for temporary protection from contamination and evaporation for samples in 96 well plates.