Learn More
Chemglass Life Sciences 2in Diameter X 1/16in Thick Borosilicate Disc
SDP
Catalog No. 50872319
Encompass_Preferred
Plate glass, disc, borosilicate, 2in Diameter X 1/16in Thick. Pyrex or Duran borosilicate glass disc. Edges on all sizes are ground and chamfered.
Catalog No. 50-872-319 Supplier Chemglass Life Sciences Supplier No. CG190642
Plate glass, disc, borosilicate, 2in Diameter X 1/16in Thick. Pyrex or Duran borosilicate glass disc. Edges on all sizes are ground and chamfered.