Learn More
Camag Scientific Tlc Plate Heater 3, 115 V, For Tlc/Hptlc Plates Up To 20 X 20 Cm, Temperature Range 25 - 200 ºc
SDP
Catalog No. 501979323
Encompass_Preferred
The Tlc Plate Heater 3 Is A Device For Heating Tlc/Hptlc Plates, Permitting Optimal Heating During The Derivatization Reaction. It Features A Ceran Heating Surface Which Is Resistant To All Common Reagents And Is Easily Cleaned. The 20 X 20 Cm Heating Surface Has A Grid To Facilitate Correct Positioning Of The Plate. Programmed And Actual Temperature Are Digitally Displayed. The Temperature Is Selectable Between 25 And 200 °C. The Plate Heater Is Protected From Overheating.
Catalog No. 50-197-9323 Supplier Camag Scientific Supplier No. 022.3307
The Tlc Plate Heater 3 Is A Device For Heating Tlc/Hptlc Plates, Permitting Optimal Heating During The Derivatization Reaction. It Features A Ceran Heating Surface Which Is Resistant To All Common Reagents And Is Easily Cleaned. The 20 X 20 Cm Heating Surface Has A Grid To Facilitate Correct Positioning Of The Plate. Programmed And Actual Temperature Are Digitally Displayed. The Temperature Is Selectable Between 25 And 200 °C. The Plate Heater Is Protected From Overheating.