Learn More
Thomas Scientific Skirted AmplateT, 96-Well, PCR, Natural
SDP
Catalog No. 501458960
Encompass_Preferred
Made of polypropylene Amplate™ Raised Rim thin wall PCR plates are the latest addition to the wide range of Simport PCR products. Offering just the right rigidity for automation, these four 96-well plates, made in a standard 8 x 12 configuration, are perfectly suited for high performance thermal cycling. Each well makes intimate contact with the heating block while quick and consistent heat transfer is ensured by a uniform wall thickness. Using a special polypropylene, samples are easily recovered thanks to a low adhesion surface. Well capacity: T323-100 and -101 Series: 250 µl, T323-103 Series: 200 µL These plates offer the right alternative to existing Robbins (-100), Corning (-101), Perkin Elmer (-103) and ABI (-104) models. T323-100 and –101 Series have a 3 mm raised rim around each tube well. T323-101 is supplied with a wide skirt extending over and under the plate on which a bar code can be affixed to facilitate identification. T323-103 will also offe
Catalog No. 50-145-8960 Supplier Thomas Scientific Supplier No. 1219F28
Made of polypropylene Amplate™ Raised Rim thin wall PCR plates are the latest addition to the wide range of Simport PCR products. Offering just the right rigidity for automation, these four 96-well plates, made in a standard 8 x 12 configuration, are perfectly suited for high performance thermal cycling. Each well makes intimate contact with the heating block while quick and consistent heat transfer is ensured by a uniform wall thickness. Using a special polypropylene, samples are easily recovered thanks to a low adhesion surface. Well capacity: T323-100 and -101 Series: 250 µl, T323-103 Series: 200 µL These plates offer the right alternative to existing Robbins (-100), Corning (-101), Perkin Elmer (-103) and ABI (-104) models. T323-100 and –101 Series have a 3 mm raised rim around each tube well. T323-101 is supplied with a wide skirt extending over and under the plate on which a bar code can be affixed to facilitate identification. T323-103 will also offe