Learn More
Chemglass Life Sciences Plate, PCR Reaction, Thin Wall, 24-Well, Green
SDP
Catalog No. 501311319
Encompass_Preferred
Plate, PCR Reaction, Thin Wall, 24-Well, Green. These PCR plates are thin-walled and designed for rapid thermal transfer. Pre-cut plates are available in 96-, 48- and 24-well. Each well has a capacity of 0.2mL. They are precision molded to ensure well-to-well and plate-to-plate uniformity. The insides of the tubes are smooth and have an inert surface which enzymes and nucleic acids do not bind. The plates will accommodate differences in expansion coefficients between the metal thermal cycler block and polypropylene tubes. On the 96-well plates, an alphanumeric grid helps with sample identification. To facilitate orientation, the bottom right corner is cut away.The AMPLATE™ is easy to seal since no cylindrical walls extend above the plate. More economical than using single tubes, it is available in four different colors or colorless. Autoclavable. Packed in re-sealable bags of 10 plates each.
Catalog No. 50-131-1319 Supplier Chemglass Life Sciences Supplier No. CLS361524G
Plate, PCR Reaction, Thin Wall, 24-Well, Green. These PCR plates are thin-walled and designed for rapid thermal transfer. Pre-cut plates are available in 96-, 48- and 24-well. Each well has a capacity of 0.2mL. They are precision molded to ensure well-to-well and plate-to-plate uniformity. The insides of the tubes are smooth and have an inert surface which enzymes and nucleic acids do not bind. The plates will accommodate differences in expansion coefficients between the metal thermal cycler block and polypropylene tubes. On the 96-well plates, an alphanumeric grid helps with sample identification. To facilitate orientation, the bottom right corner is cut away.The AMPLATE™ is easy to seal since no cylindrical walls extend above the plate. More economical than using single tubes, it is available in four different colors or colorless. Autoclavable. Packed in re-sealable bags of 10 plates each.