Learn More
VWR INTERNATIONAL PLATE PCR SEMISKIRT NAT CS100
Catalog No. NC0684948
Encompass
Pcr Plates From Sorenson Bioscience Contain Bold-Printed Alphanumerics And Fit All Leading Thermal Cyclers, Dna Sequencing/Detection Instruments, And Real-Time Pcr Instruments. Tight Manufacturing Tolerances And Sbs Standard Dimensions Create Plates Ideal For Automated Processes. Volume 0.2 Ml; Natural; Case Of 100
Catalog No. NC0684948 Supplier VWR INTERNATIONAL Supplier No. 14216160
Pcr Plates From Sorenson Bioscience Contain Bold-Printed Alphanumerics And Fit All Leading Thermal Cyclers, Dna Sequencing/Detection Instruments, And Real-Time Pcr Instruments. Tight Manufacturing Tolerances And Sbs Standard Dimensions Create Plates Ideal For Automated Processes. Volume 0.2 Ml; Natural; Case Of 100