Learn More
Andwin Scientific PLATE ROLLER..
SDP
Catalog No. NC0081836
Encompass_Preferred
The Plate Roller guarantees secure, tight, uniform seal of adhesive films and foils to microplate wells. This product has an overall length of 10.2 centimeters of 4 inches.
Catalog No. NC0081836 Supplier Andwin Scientific Supplier No. 60941118
The Plate Roller guarantees secure, tight, uniform seal of adhesive films and foils to microplate wells. This product has an overall length of 10.2 centimeters of 4 inches.