Learn More
Nacalai USA Inc PLATE SEAL EPS 1 BOX
Catalog No. 501037301
Encompass_Preferred
Easy Piercing Seal (EPS) from BioChromato punctures easily to prevent needle clogging. In addition, no adhesive residues are left on the plate or injector needle. The EPS plate seal is highly resistant to organic solvents.
Catalog No. 50-103-7301 Supplier Nacalai USA Inc Supplier No. R37.001.00CS
Easy Piercing Seal (EPS) from BioChromato punctures easily to prevent needle clogging. In addition, no adhesive residues are left on the plate or injector needle. The EPS plate seal is highly resistant to organic solvents.