Learn More
Andwin Scientific Brayer Roller, 4inch Soft Rubber, for uniformly sealing film to microplates
SDP
Catalog No. NC1664415
Encompass_Preferred
Brayer Roller, 4inch Soft Rubber, for uniformly sealing film to microplates
Catalog No. NC1664415 Supplier Andwin Scientific Supplier No. BRAY4000
Brayer Roller, 4inch Soft Rubber, for uniformly sealing film to microplates