Learn More
Sigma Aldrich Fine Chemicals Biosciences Greiner multiwell plate sealers, 100/pk
Catalog No. NC1679818
Encompass
Greiner microplate sealers are RNase free, DNase free and nonpyrogenic. They feature a temperature tolerance range of -70 °C up to 110 °C. VIEWseal™ sealing film, pressure sensitive adhesive, transparent, double-perforated sides.
Catalog No. NC1679818 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z617571100EA
Greiner microplate sealers are RNase free, DNase free and nonpyrogenic. They feature a temperature tolerance range of -70 °C up to 110 °C. VIEWseal™ sealing film, pressure sensitive adhesive, transparent, double-perforated sides.