Learn More
Thomas Scientific AlumaSeal CS Foil, -80øC
SDP
Catalog No. 501458237
Encompass
Specially formulated for cold storage to -80°C Excellent barrier properties to slow down evaporation Easily pierceable with pipet tips or robotic probes for sample recovery Available non-sterile and sterile AlumaSeal CS sealing films are specially formulated with an adhesive that withstands cold storage at temperatures to -80°C. Unlike other sealing films in the AlumaSeal group, AlumaSeal CS sealing films are not recommended for PCR or thermal cycling. A single non-perforated end tab simplifies application. Simply hold the tab and strip the backing from the body of the sealing film as it lays on the plate to avoid curling. Dimensions are132.6 x 82.6 mm, including the single 9.5 mm end tab. Recommended temperature range: -80°C to ambient. AlumaSeal CS sealing films are certified DNase-, RNase-, and nucleic-acid-free. Excel Scientific Sealing Film Selection Chart
Catalog No. 50-145-8237 Supplier Thomas Scientific Supplier No. 1230Y24
Specially formulated for cold storage to -80°C Excellent barrier properties to slow down evaporation Easily pierceable with pipet tips or robotic probes for sample recovery Available non-sterile and sterile AlumaSeal CS sealing films are specially formulated with an adhesive that withstands cold storage at temperatures to -80°C. Unlike other sealing films in the AlumaSeal group, AlumaSeal CS sealing films are not recommended for PCR or thermal cycling. A single non-perforated end tab simplifies application. Simply hold the tab and strip the backing from the body of the sealing film as it lays on the plate to avoid curling. Dimensions are132.6 x 82.6 mm, including the single 9.5 mm end tab. Recommended temperature range: -80°C to ambient. AlumaSeal CS sealing films are certified DNase-, RNase-, and nucleic-acid-free. Excel Scientific Sealing Film Selection Chart