Learn More
Phenomenex Inc TLC Plates Silica
Catalog No. 502412867 Shop All Phenomenex Products
Encompass_Preferred
TLC Plates Silica Claricep TLC Plates Silica, 25×75mm; 60A; Glass backing; F254
Catalog No. 50-241-2867 Supplier Phenomenex Inc Supplier No. TCSF75250
TLC Plates Silica Claricep TLC Plates Silica, 25×75mm; 60A; Glass backing; F254