Learn More
Grainger Acorn Controls Orifice Plate Spring, Stainless Steel, 3/8 in. H, 10/Pk.
Catalog No. 502785175
Encompass_Preferred
This orifice plate spring is a plumbing component designed for use with Air-Trol valves and wash fountain systems. It serves as a direct replacement part constructed for durability in maintenance applications.
  • Constructed from stainless steel
  • Compatible with Air-Trol valves and series wash fountain equipment
  • Measures 3/8 in. H x 1/4 in. dia.
  • Packaged in a quantity of 10
Catalog No. 50-278-5175 Supplier Grainger Supplier No. 19C793
This orifice plate spring is a plumbing component designed for use with Air-Trol valves and wash fountain systems. It serves as a direct replacement part constructed for durability in maintenance applications.
  • Constructed from stainless steel
  • Compatible with Air-Trol valves and series wash fountain equipment
  • Measures 3/8 in. H x 1/4 in. dia.
  • Packaged in a quantity of 10