Description
- Stainless steel plate stand with anti-slip silicone mat
- Drive unit height adjustable allowing for hands free work
- Vessel Holder
- Safety Positioning ring: safety feature to prevent uncontrolled adjustment of drive unit
Specifications
Specifications
| Description | Stand for PT 1200E with height-adjustable holder for the drive unit and fastening the base station. |
| For Use With (Equipment) | Polytron™ PT 1200 E |
| Includes | Silicone Anti-Slip Mat |
| Model | ST-P 13/320 |
| Product Type | Plate Stand |