Learn More
Research Products International Corp Plate Stand for Well Orienter, with Rubber Feet and Neoprene Backing to Prevent Slipping
SDP
Catalog No. 50998941
Encompass
The Well Orienter System increases accuracy and efficiency with 96 well micro-plate procedures. The plate stand holds an index card and micro-plate at the optimum viewing angle to view for pipetting while sitting or standing. Stand has rubber feet and a neoprene backing to prevent slipping.
Catalog No. 50-998-941 Supplier Research Products International Corp Supplier No. 248805
The Well Orienter System increases accuracy and efficiency with 96 well micro-plate procedures. The plate stand holds an index card and micro-plate at the optimum viewing angle to view for pipetting while sitting or standing. Stand has rubber feet and a neoprene backing to prevent slipping.