Learn More
Med Vet International Standard TPLO Plate, 3.5mm, Left, J0438, Each
Supplier:  Med Vet International JOR438
Encompass_Preferred
JorVet version of the original TPLO plates.Made of 316LVM implant grade stainless steelMilled, not cast The JorVet plate has been modified in the head region to make contouring easy while retaining the stiffness of 316LVM elsewhere for stability and rapid healing. The lower profile makes closure easier.
Catalog No. 50-005-82354