Learn More
Cole-Parmer Argos Technologies Sterile Tissue Culture 12- Well Plate, 100/cs
Catalog No. 501960634 Shop All Cole-Parmer Products
Encompass_Preferred
Use these plates to serve various cell and tissue culture needs. The plates are manufactured in a cleanroom environment for optimal quality. They have treated surfaces of an exclusive coating, which causes the surface to become negatively charged and hydrophilic, allowing for consistent and superior cell attachment and growth. Plates have individual condensation rings and a one-way fitting lid to reduce cross-contamination. The alphanumeric notation on each well allows for simple identification.
Catalog No. 50-196-0634 Supplier Cole-Parmer Supplier No. 0192772
Use these plates to serve various cell and tissue culture needs. The plates are manufactured in a cleanroom environment for optimal quality. They have treated surfaces of an exclusive coating, which causes the surface to become negatively charged and hydrophilic, allowing for consistent and superior cell attachment and growth. Plates have individual condensation rings and a one-way fitting lid to reduce cross-contamination. The alphanumeric notation on each well allows for simple identification.