Learn More
Cole-Parmer Argos Technologies Sterile Tissue Culture 96-Well Plate, 100/cs
Catalog No. 501960587 Shop All Cole-Parmer Products
Encompass
Use these culture well plates to serve your various cell and tissue culture needs. The plates are manufactured in a cleanroom environment for optimal quality. They have treated surfaces of an exclusive coating, which causes the surface to become negatively charged and hydrophilic, allowing for consistent and superior cell attachment and growth. They also have individual condensation rings, and a one-way fitting lid to reduce cross-contamination. The alphanumeric notation on each well allows for simple identification.
Catalog No. 50-196-0587 Supplier Cole-Parmer Supplier No. 0192778
Use these culture well plates to serve your various cell and tissue culture needs. The plates are manufactured in a cleanroom environment for optimal quality. They have treated surfaces of an exclusive coating, which causes the surface to become negatively charged and hydrophilic, allowing for consistent and superior cell attachment and growth. They also have individual condensation rings, and a one-way fitting lid to reduce cross-contamination. The alphanumeric notation on each well allows for simple identification.