Learn More
IPHASE PHARMA SERVICES LLC Plateable ICR/CD-1 Mouse Hepatocytes,Male,5million
Catalog No. NC3966932
Encompass_Preferred
Primary Hepatocytes are isolated from human and animal liver tissues. It's a tool for cytochrome P450 induction, Hepatotoxicity assays and siRNA devlivery, Transfection, Sandwitch model, 3D Primary Hepatocyte Spheroid Model (Three-Dimensional Liver Culture system) and organoids culture. IPHASE provides both suspension and plateable Hepatocytes, animal or human and speical species includes: cat(feline), Guinea pig, BALB/c Nude, Ferret, Gerbil customize hepatocytes. Plateable Hepatocytes need to be used in conjunction with Thaw medium, Plateable medium, Maintenance medium and Collagen Coated Plate.These products require shipment in LN2 container therefore customer need to return the container.
Catalog No. NC3966932 Supplier IPHASE PHARMA SERVICES LLC Supplier No. 01932E1.21
Primary Hepatocytes are isolated from human and animal liver tissues. It's a tool for cytochrome P450 induction, Hepatotoxicity assays and siRNA devlivery, Transfection, Sandwitch model, 3D Primary Hepatocyte Spheroid Model (Three-Dimensional Liver Culture system) and organoids culture. IPHASE provides both suspension and plateable Hepatocytes, animal or human and speical species includes: cat(feline), Guinea pig, BALB/c Nude, Ferret, Gerbil customize hepatocytes. Plateable Hepatocytes need to be used in conjunction with Thaw medium, Plateable medium, Maintenance medium and Collagen Coated Plate.These products require shipment in LN2 container therefore customer need to return the container.