Learn More
Boca Scientific Inc PlateBlock™, 125ml
Catalog No. NC1894474
Encompass
PlateBlock™ is a blocking reagent optimized for serological assays (antigen-down assays) that greatly improves the reliability of the assay. It does not contain any protein components and is produced without any raw materials of animal origin. Problems with blocking the microplate surface occur frequently in serological ELISA and often lead to false positives. PlateBlock™ has been specifically designed to overcome these challenges. By saturating plastic surfaces, PlateBlock™ prevents non-specific binding, cross reactivity between the blocking surface and the detector antibody, and reduces background. Incorporating PlateBlock™ into your serological ELISA will greatly improve the sensitivity of the assay and prevent false positives and negatives in antibody tests.
Catalog No. NC1894474 Supplier Boca Scientific Inc Supplier No. 112 125
Add to Cart
Add to Cart
PlateBlock™ is a blocking reagent optimized for serological assays (antigen-down assays) that greatly improves the reliability of the assay. It does not contain any protein components and is produced without any raw materials of animal origin. Problems with blocking the microplate surface occur frequently in serological ELISA and often lead to false positives. PlateBlock™ has been specifically designed to overcome these challenges. By saturating plastic surfaces, PlateBlock™ prevents non-specific binding, cross reactivity between the blocking surface and the detector antibody, and reduces background. Incorporating PlateBlock™ into your serological ELISA will greatly improve the sensitivity of the assay and prevent false positives and negatives in antibody tests.