Learn More
Research Products International Corp PlateFuge Microplate MicroCentrifuge for DeepWell and PCR Plates, 230 volt
SDP
Catalog No. 501977010
Encompass
The first "mini" sized centrifuge with a swing out rotor for microplates. Centrifuging plates before PCR ensures all reactants are in the bottom of the wells for proper concentrations and improved yields.
In spite of its remarkably small footprint (9x10 in.), the PlateFuge includes a uniquely designed, swing out rotor (patent pending) capable of securing 2 microplates. The easily accessible rotor chamber includes two plate carriers that rest at a 75° angle. This allows plates to be inserted confidently without sealing tapes or caps. Upon closing the lid of the centrifuge, the rotor accelerates and the centrifugal force "swings" the plates into a horizontal position. Any droplets on the walls of the plate quickly concentrate into the well bottoms.
Catalog No. 50-197-7010 Supplier Research Products International Corp Supplier No. 112170230
The first "mini" sized centrifuge with a swing out rotor for microplates. Centrifuging plates before PCR ensures all reactants are in the bottom of the wells for proper concentrations and improved yields.
In spite of its remarkably small footprint (9x10 in.), the PlateFuge includes a uniquely designed, swing out rotor (patent pending) capable of securing 2 microplates. The easily accessible rotor chamber includes two plate carriers that rest at a 75° angle. This allows plates to be inserted confidently without sealing tapes or caps. Upon closing the lid of the centrifuge, the rotor accelerates and the centrifugal force "swings" the plates into a horizontal position. Any droplets on the walls of the plate quickly concentrate into the well bottoms.