Learn More
Medchemexpress LLC Platelet-activating factor acetylhydrolases 1b2 and 1b3 inhibitor | 942285-55-4 | 99.69% | 503.65 | 1 ML
SDP
Catalog No. 5000343605
Encompass_Preferred
P11 is a selective inhibitor of platelet-activating factor acetylhydrolases (PAFAHs) 1b2 and 1b3 that has been shown to impair cancer cell survival. It exhibits IC50 values of approximately 40 nM for PAFAH1b2 and 900 nM for PAFAH1b3.
  • Selective inhibitor of PAFAH1b2 and PAFAH1b3.
  • Impairs cancer cell survival.
  • High purity (99.69%).
  • Available in various quantities and as a 10 mM solution in DMSO.
  • Well-characterized with available data sheet, COA, and SDS.
Catalog No. 50-003-43605 Supplier Medchemexpress LLC Supplier No. HY1110531MG
P11 is a selective inhibitor of platelet-activating factor acetylhydrolases (PAFAHs) 1b2 and 1b3 that has been shown to impair cancer cell survival. It exhibits IC50 values of approximately 40 nM for PAFAH1b2 and 900 nM for PAFAH1b3.
  • Selective inhibitor of PAFAH1b2 and PAFAH1b3.
  • Impairs cancer cell survival.
  • High purity (99.69%).
  • Available in various quantities and as a 10 mM solution in DMSO.
  • Well-characterized with available data sheet, COA, and SDS.