Learn More
Creative Biomart Platelet-Derived Endothelial Cell Growth Factor (PD-ECGF)
Catalog No. 502084274
Encompass_Preferred
Platelet-derived endothelial cell growth factor is a 45 kDa endothelial cell mitogen of pI 4.0-4.8 originally purified from human platelets. PD-ECGF is chemotactic for bovine aortic endothelial cells, however it does not induce smooth muscle cell migration. Potency: 5-30 ng/mL EC50
Catalog No. 50-208-4274 Supplier Creative Biomart Supplier No. THP0223
Platelet-derived endothelial cell growth factor is a 45 kDa endothelial cell mitogen of pI 4.0-4.8 originally purified from human platelets. PD-ECGF is chemotactic for bovine aortic endothelial cells, however it does not induce smooth muscle cell migration. Potency: 5-30 ng/mL EC50