Learn More
STEMCELL Technologies Human Recombinant PDGF-AB, Size: 1000 µg
SDP
Supplier:  STEMCELL Technologies 78096.2
Encompass
Platelet-derived growth factor (PDGF) is a dimeric glycoprotein consisting of two disulfide bridge-stabilized polypeptide chains, A and B, which are assembled as heterodimers (PDGF-AB) or homodimers (PDGF-AA and PDGF-BB) (Fretto et al.; Westermark & Heldin). PDGF signals through the receptor tyrosine kinases PDGFRalpha and PDGFRbeta. It has been shown that PDGF-induced migration involves signaling pathways involving MEK/ERK, EGFR, Src, and PI3K/AKT (Kim et al.). PDGF is a potent mitogen for cells of mesenchymal origin- like fibroblasts, glial cells, and vascular smooth muscle cells. PDGF has been implicated in pathogenesis of atherosclerosis, glomerulonephritis, cancer, and in the contraction of vascular smooth muscle cells of rat aortic tissues (Fretto et al.; Sachinidis et al.). It has been shown that PDGF-AB together with 5-Azacytidine (Catalog #72012), induces the conversion of mature bone and fat cells into tissue-regenerative multipotent stem cells (Chandrakanthan et al.).
Catalog No. 50-226-9658