Learn More
Creative Biomart Platelet-Derived Growth Factor-AA (PDGFAA)
Catalog No. 502084275
Encompass_Preferred
PDGF exists both in homo and heterodimeric forms and stimulates and -cell surface receptors. Vascular endothelial growth factor expression is regulated by PDGF-AA autocrine signalling and is known to promote tumor angiogenesis in vivo. Potency: 1.0-3.0 ng/mL ED50.
Catalog No. 50-208-4275 Supplier Creative Biomart Supplier No. THP0224
PDGF exists both in homo and heterodimeric forms and stimulates and -cell surface receptors. Vascular endothelial growth factor expression is regulated by PDGF-AA autocrine signalling and is known to promote tumor angiogenesis in vivo. Potency: 1.0-3.0 ng/mL ED50.