Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Platelet-derived growth factor C (PDGF-CC) | 50 UG
SDP

Catalog No. 5000291568
Encompass_Preferred

Recombinant Fc-tagged platelet-derived growth factor C (PDGF-CC) derived from cynomolgus monkey and expressed in HEK293 cells. Supplied as a lyophilized powder for research use, it is intended for cell proliferation and signaling assays involving mesenchymal cells, angiogenesis, and wound healing studies.

  • Species: Cynomolgus.
  • Expression system: HEK293 cells.
  • Tag: human Fc.
  • Formulation: lyophilized from PBS with protectants (trehalose/mannitol, Tween 80).
  • Molecular weight: ~45-50 kDa (SDS-PAGE).
  • Endotoxin: <1 EU/μg.
  • Biological activity: ED50 15.09 ng/mL in NIH/3T3 proliferation assay.
  • Storage: store lyophilized at -20°C; aliquot and store at -80°C for long term.

Catalog No. 50-002-91568 Supplier Medchemexpress LLC Supplier No. HYP7464050UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant Fc-tagged platelet-derived growth factor C (PDGF-CC) derived from cynomolgus monkey and expressed in HEK293 cells. Supplied as a lyophilized powder for research use, it is intended for cell proliferation and signaling assays involving mesenchymal cells, angiogenesis, and wound healing studies.

  • Species: Cynomolgus.
  • Expression system: HEK293 cells.
  • Tag: human Fc.
  • Formulation: lyophilized from PBS with protectants (trehalose/mannitol, Tween 80).
  • Molecular weight: ~45-50 kDa (SDS-PAGE).
  • Endotoxin: <1 EU/μg.
  • Biological activity: ED50 15.09 ng/mL in NIH/3T3 proliferation assay.
  • Storage: store lyophilized at -20°C; aliquot and store at -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.