Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Platelet derived growth factor C (PDGF-CC), cynomolgus hFc fusion protein | 10 UG
SDP

Catalog No. 5000293353
Encompass_Preferred

Recombinant PDGF-CC protein derived from cynomolgus and fused to a human Fc domain, expressed in HEK293 cells and supplied as a lyophilized powder. The protein is formulated for stability and demonstrates biological activity in cell proliferation assays; see manufacturer datasheet for detailed handling, reconstitution, and storage instructions.

  • Recombinant cynomolgus PDGF-CC expressed in HEK293 cells.
  • Human Fc fusion (hFc) for increased stability and detection.
  • Lyophilized powder formulated in PBS with protectants (trehalose/mannitol and 0.01% Tween 80).
  • Endotoxin <1 EU/μg as determined by LAL.
  • Biological activity ED50 = 15.09 ng/mL in NIH/3T3 proliferation assay.
  • Molecular weight ~45-50 kDa by SDS-PAGE under reducing conditions.
  • Store at -20°C; reconstitute to ≥100 μg/mL; freeze aliquots for long-term storage.

Catalog No. 50-002-93353 Supplier Medchemexpress LLC Supplier No. HYP7464010UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant PDGF-CC protein derived from cynomolgus and fused to a human Fc domain, expressed in HEK293 cells and supplied as a lyophilized powder. The protein is formulated for stability and demonstrates biological activity in cell proliferation assays; see manufacturer datasheet for detailed handling, reconstitution, and storage instructions.

  • Recombinant cynomolgus PDGF-CC expressed in HEK293 cells.
  • Human Fc fusion (hFc) for increased stability and detection.
  • Lyophilized powder formulated in PBS with protectants (trehalose/mannitol and 0.01% Tween 80).
  • Endotoxin <1 EU/μg as determined by LAL.
  • Biological activity ED50 = 15.09 ng/mL in NIH/3T3 proliferation assay.
  • Molecular weight ~45-50 kDa by SDS-PAGE under reducing conditions.
  • Store at -20°C; reconstitute to ≥100 μg/mL; freeze aliquots for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.