Learn More
Medchemexpress LLC Platelet-derived growth factor subunit B (PDGF-BB), cynomolgus | 5 UG
SDP
Catalog No. 5000291152
Encompass_Preferred
Recombinant PDGF-BB (cynomolgus) expressed in HEK293 cells and fused to a mouse Fc tag, provided as a sterile, 0.22 μm filtered PBS solution (pH 7.4) for use in cell proliferation and functional assays. Activity is measured by ED50 in Balb/c 3T3 proliferation assays; aliquot and store frozen to preserve activity.
Catalog No. 50-002-91152 Supplier Medchemexpress LLC Supplier No. HYP759665UG
Recombinant PDGF-BB (cynomolgus) expressed in HEK293 cells and fused to a mouse Fc tag, provided as a sterile, 0.22 μm filtered PBS solution (pH 7.4) for use in cell proliferation and functional assays. Activity is measured by ED50 in Balb/c 3T3 proliferation assays; aliquot and store frozen to preserve activity.