Learn More
Creative Biomart Platelet Factor-4 (PF4) Protein
Catalog No. 502084282
Encompass_Preferred
PF4 Human Recombinant produced in E. Coli is a single, non-glycosylated polypeptide chain containing 70 amino acids and having a molecular mass of 7.8 kDa.
Catalog No. 50-208-4282 Supplier Creative Biomart Supplier No. THP0231
PF4 Human Recombinant produced in E. Coli is a single, non-glycosylated polypeptide chain containing 70 amino acids and having a molecular mass of 7.8 kDa.