Learn More
Verax Biomedical™ Platelet PGDprime™ Controls
Catalog No. 23051013
Change view
Click to view available options
Quantity:
30 Tests
Catalog No. Quantity
23-051-013 30 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-051-013 Supplier Verax Biomedical™ Supplier No. PRM30C
Add to Cart
Add to Cart
This item is not returnable. View return policy
Positive and Negative controls to verify the performance of the Platelet PGDprime test.
  • 1 bottle of each control per package
  • Contains bacterial antigens
  • Only for use with the Platelet PGDprime test
  • Ready to use in dropper bottles
  • Sufficient volume for 30 QC runs
Specifications
CE Marker No
Packaging Type Box
Quantity 30 Tests