Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Verax Biomedical™ Platelet PGDprime™ Test

Catalog No. 23051017
Change view
Click to view available options
Quantity:
100 Tests
20 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-051-017 20 Tests
23-051-010 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-051-017 Supplier Verax Biomedical™ Supplier No. PRM20
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapid test for the detection of Gram-positive and Gram-negative bacterial contamination in platelets. Allows extension of platelet dating from 5 to 7 days when testing apheresis platelets in plasma.

  • Easy to perform qualitative test
  • Detects both aerobes and anaerobes
  • Observed specificity: 100% / LCL: 99.9%
  • 150 μL sample size
  • No centrifugation or precision pipetting required
  • Includes reagents and disposables required to perform the test
  • Ready to use liquid reagents in dropper bottles
  • Colored line in read window indicates a reactive result
  • Built in procedural control verifies assay validity
  • FDA cleared for use with: Leukoreduced apheresis platelets in plasma or PAS-C, polls of up to 6 pre-storage or post-storage pooled WBD platelets and individual WBD platelets
  • CLIA Complexitiy: Moderate
  • Ready to use liquid reagents in dropper bottles
  • Colored line in read window indicates a reactive result

Specifications

CE Marker No
Packaging Type Box
Quantity 20 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.