Learn More
Cayman Chemical Platelet-Derived Growth Factor Receptors α and β Reporter Assay System, 96 Well
SDP
Catalog No. 502913609
Encompass_Preferred
This Human Platelet-Derived Growth Factor Receptors α and β (PDGFRα/β) Reporter Assay Kit is an all-inclusive firefly luciferase reporter assay system that includes in addition to PDGFRα/β reporter cells
Catalog No. 50-291-3609 Supplier Cayman Chemical Supplier No. 3669796 WELL
This Human Platelet-Derived Growth Factor Receptors α and β (PDGFRα/β) Reporter Assay Kit is an all-inclusive firefly luciferase reporter assay system that includes in addition to PDGFRα/β reporter cells