Learn More
Cayman Chemical Platelet-Derived Growth Factor Receptors α and β Reporter Assay System, 3x32 Well
SDP
Catalog No. 502913608
Encompass_Preferred
This Human Platelet-Derived Growth Factor Receptors α and β (PDGFRα/β) Reporter Assay Kit is an all-inclusive firefly luciferase reporter assay system that includes in addition to PDGFRα/β reporter cells
Catalog No. 50-291-3608 Supplier Cayman Chemical Supplier No. 366973X32 WELL
This Human Platelet-Derived Growth Factor Receptors α and β (PDGFRα/β) Reporter Assay Kit is an all-inclusive firefly luciferase reporter assay system that includes in addition to PDGFRα/β reporter cells