Learn More
MilliporeSigma™ Supelco™ PlatePrep 96-well Vacuum Manifold
Catalog No. 111003078 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
96-well Vacuum Manifold Manifold only
Catalog No.
11-100-3078 96-well Vacuum Manifold Manifold only
1 options
Catalog No. 11-100-3078 Supplier MilliporeSigma™ Supelco™ Supplier No. 57192U
Add to Cart
Add to Cart
The PlatePrep vacuum manifold consists of a clear acrylic top allowing for easier inspection of flow rates during SPE 96-well plate processing. The polypropylene base offers excellent chemical resistance while a single remote vacuum gauge/bleed valve controls flow through all the wells.
Use this compact vacuum manifold in conjunction with a Discovery SPE 96-well plate to process up to 96 samples concurrently. The single valve control, parallel processing capabilities, and uniform flow dynamics allow for easier method development, reduce clutter, and improve reproducibility. Unused wells can be covered and used at a later date.
A PlatePrep 96-Well Starter Kit (Cat. No. 575650-U) is available to analysts new to 96-well SPE technology. Included with the Starter Kit is PlatePrep Vacuum Manifold and sample units of key accessory items necessary for 96-well SPE.
TRUSTED_SUSTAINABILITY
Specifications
Description 96-well μVacuum Manifold Manifold only