Learn More
Mitegen Plates, CBT Crystallization, Case Of 40
SDP
Catalog No. 501492102
Encompass_Preferred
The CombiClover 1500 Crystallization plates offer the exact same design and utility as our CombiClover 500 crystallization plates, but offer larger capacity in the reservoir and drop chambers. A central reservoir is connected to four satellite drop chambers via dedicated vapor diffusion channels to create a novel combinatorial "crystallization clover". but offer larger capacity in the reservoir and drop chambers.-Plate dimensions are 112.0mm x 165.0mm x 24.0mm-The central reservoir volume is 1.5 millilters-The satellite drop chamber volume is 50 microliters
Catalog No. 50-149-2102 Supplier Mitegen Supplier No. 1009544
The CombiClover 1500 Crystallization plates offer the exact same design and utility as our CombiClover 500 crystallization plates, but offer larger capacity in the reservoir and drop chambers. A central reservoir is connected to four satellite drop chambers via dedicated vapor diffusion channels to create a novel combinatorial "crystallization clover". but offer larger capacity in the reservoir and drop chambers.-Plate dimensions are 112.0mm x 165.0mm x 24.0mm-The central reservoir volume is 1.5 millilters-The satellite drop chamber volume is 50 microliters