Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces
GSA_VA

Catalog No. 1256714 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Black
Clear
White
Volume (Metric) Working:
100 μL
Product Type:
Microplate
Solid Module F8 (x12) in Frame
Solid plate 96-Well
Surface Treatment:
Amine
Covalent Binding
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Volume (Metric) Working Product Type Surface Treatment
12-567-14 Clear 100 μL Microplate Amine
12-567-12 Clear 100 μL Microplate Amine
12-567-10 White 100 μL Solid plate 96-Well Covalent Binding
12-567-11 Black 100 μL Microplate Amine
12-567-13 Clear 100 μL Microplate Amine
12-565-132 Clear 100 μL Solid Module F8 (x12) in Frame Covalent Binding
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 12-567-14 Supplier Thermo Scientific™ Supplier No. 436023
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reduce the amount of reagents used in the lab by using these plates and modules with covalent binding surfaces.

Reduce the amount of reagents used in the lab with Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces.

Covalent Surfaces (Thermo Scientific™ CovaLink™ and Immobilizer Amino)

  • Minimal leaching
  • Withstands vigorous washing
  • Coating with lower amounts of reagent may be possible
  • Control of orientation

Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Breakapart Breakable Strips
Detection Method Colorimetric
Packaging 30 Cases
Binding Type Covalent
Color Clear
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 350 μL
Volume (Metric) Working 100 μL
Sterility Non-sterile
Surface Treatment Amine
Total Volume (Metric) Well 350 μL
Well Shape Round, C Bottom
No. per Case 30
No. per Pack 5
Plate Blocking Unspecified
Target Molecule Peptides
Show More Show Less
Which type of Immobilizer Amino plates and strips should I use for the different detection methods?

We recommend using transparent polystyrene plates and strips for colorimetric assays, white opaque polystyrene plates and strips for bio and chemi-luminescence assays, and black opaque polystyrene plates and strips for fluorescence assays.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.

What advantages do your covalent binding surfaces offer over passive surfaces?

While adsorption of biomolecules to our passive surfaces is more electrostatic in nature, our CovaLink and Immobilizer Amino surfaces present an option for true covalent bonding of molecules with specific free residues. CovaLink possesses a secondary amine structure, which will covalently bond to biomolecules with a free carboxyl or phosphate group. This allows for a much more robust and specific coating, which will often tolerate more rigorous washing steps and eliminate the need for blocking.

Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.