Learn More
Mitegen Plates, XJR Crystallization, Case Of 40
SDP
Catalog No. 501492104
Encompass_Preferred
The Compact 300 Crystallization plate utilizes our patented sitting drop well design and vapor diffusion channel. This plate has 96 reservoirs with 96 drop chambers.-Plate dimensions are 85.5mm x 127.8mm x 14.4mm-Plates are compatible with SBS (Society for Biomolecular Screening) standard laboratory automated instrumentation.-Drop chambers are 9mm apart in both horizontal and vertical directions for eay use with multi-channel pipettors.-The reservoir volume is 300 microliters.-The drop chamber volume is 6 microliters.
Catalog No. 50-149-2104 Supplier Mitegen Supplier No. 1009546
The Compact 300 Crystallization plate utilizes our patented sitting drop well design and vapor diffusion channel. This plate has 96 reservoirs with 96 drop chambers.-Plate dimensions are 85.5mm x 127.8mm x 14.4mm-Plates are compatible with SBS (Society for Biomolecular Screening) standard laboratory automated instrumentation.-Drop chambers are 9mm apart in both horizontal and vertical directions for eay use with multi-channel pipettors.-The reservoir volume is 300 microliters.-The drop chamber volume is 6 microliters.