Learn More
Mitegen Plates, XPT Crystallization, Case Of 10
SDP
Catalog No. 501492105
Encompass_Preferred
The CombiClover 4 Chamber Crystallization plate utilizes four partitioned reservoir chambers in the place of one central reservoir. These four partitioned chambers are each connected to a satellite drop chamber via dedicated vapor diffusion channels to create a variation of the novel combinatorial "crystallization clover" provided in the plates. Each plate has 24 of the partitioned reservoirs and 96 flat-bottom sitting drop chambers. This design allows four different experiments to be conducted in each clover, with each portion containing a different crystallany, and allowing them to be investigated simultaneously.-Plate dimensions are 112.0mm x 165.0mm x 24.0mm-The capacity of each of the four partitioned areas of the reservoir is 250 microliters-The satellite drop chamber volume is 50 microliters.
Catalog No. 50-149-2105 Supplier Mitegen Supplier No. 1009547
The CombiClover 4 Chamber Crystallization plate utilizes four partitioned reservoir chambers in the place of one central reservoir. These four partitioned chambers are each connected to a satellite drop chamber via dedicated vapor diffusion channels to create a variation of the novel combinatorial "crystallization clover" provided in the plates. Each plate has 24 of the partitioned reservoirs and 96 flat-bottom sitting drop chambers. This design allows four different experiments to be conducted in each clover, with each portion containing a different crystallany, and allowing them to be investigated simultaneously.-Plate dimensions are 112.0mm x 165.0mm x 24.0mm-The capacity of each of the four partitioned areas of the reservoir is 250 microliters-The satellite drop chamber volume is 50 microliters.