Learn More
Cole-Parmer Speirs Robertson AMS-40x50 AMS Balance Platform, Sorbothane Isolators; 40x50 cm
Supplier:  Cole-Parmer 1150005
Encompass
The AMS models use sorbothane isolators, which is perfect for heavy instruments. With a natural frequency of 7 Hz, the majority of building vibrations will be removed. This platform removes up to 90 percent of vibrations to improve performance of balances. Combination of isolators and sound-deadened layer construction removes both building and surface vibrations. Seamless pharma-grade (Ra finish 0.3) stainless steel worktops are chemical resistant and meet stringent cleaning requirements.
Catalog No. NC0939163