Learn More
Thomas Scientific Shaking platform predrilled for flask clamps
SDP
Catalog No. 501460158
Encompass
The 311DS is a high performance incubator with a built-in orbital shaker offers stable temperature conditions. A broad temperature range expands the use of this incubator to liquid cultures, washing blots and other shaking applications requiring strict temperature control. Outstanding uniformity and accuracy Gasket door with large viewing area Wide selection of accessories Broad temperature range Adjustable shelves Heat Stress Protection Temperature within the 311DS is precisely regulated by the SmartChek™ temperature control system. All electronics are located away from the chamber to protect them from heat related stress. The sturdy, integral shaker has a 19 mm orbit. A flat platform with non-slip rubber mat, as well as a variety of flask platforms are available separately. Platforms can hold as many as four 2L and six 1L flasks. The gasket lined door has a large, integral viewing area for checking samples without disturbing the chamber contents. Opening the door stops operatio
Catalog No. 50-146-0158 Supplier Thomas Scientific Supplier No. 1186X53
Add to Cart
Add to Cart
The 311DS is a high performance incubator with a built-in orbital shaker offers stable temperature conditions. A broad temperature range expands the use of this incubator to liquid cultures, washing blots and other shaking applications requiring strict temperature control. Outstanding uniformity and accuracy Gasket door with large viewing area Wide selection of accessories Broad temperature range Adjustable shelves Heat Stress Protection Temperature within the 311DS is precisely regulated by the SmartChek™ temperature control system. All electronics are located away from the chamber to protect them from heat related stress. The sturdy, integral shaker has a 19 mm orbit. A flat platform with non-slip rubber mat, as well as a variety of flask platforms are available separately. Platforms can hold as many as four 2L and six 1L flasks. The gasket lined door has a large, integral viewing area for checking samples without disturbing the chamber contents. Opening the door stops operatio