Learn More
Grainger Platform Holds 6 Standard Micro Plates
Catalog No. 502749516
Encompass_Preferred
The Platform Holds 6 Standard Microplates is a laboratory equipment product made by Thermo Scientific. It is designed to hold up to 6 standard 96-well microplates in an organized stack for easy access and identification. The platform is constructed of acrylic and has an overall dimension of 5-1/4" length x 3-3/8" width x 3-3/4" height. It can accommodate both skirted and non-skirted microplates. The platform features alpha-numeric grid references for easy plate location. This product provides an organized storage solution for standard microplates in laboratory settings.
Catalog No. 50-274-9516 Supplier Grainger Supplier No. 14N133
The Platform Holds 6 Standard Microplates is a laboratory equipment product made by Thermo Scientific. It is designed to hold up to 6 standard 96-well microplates in an organized stack for easy access and identification. The platform is constructed of acrylic and has an overall dimension of 5-1/4" length x 3-3/8" width x 3-3/4" height. It can accommodate both skirted and non-skirted microplates. The platform features alpha-numeric grid references for easy plate location. This product provides an organized storage solution for standard microplates in laboratory settings.