Learn More
Fisherbrand™ Platform Mats for Rockers and Wavers
Catalog No. 02217735 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Rockers
Catalog No. For Use With (Equipment)
02-217-735 Rockers
1 options
Catalog No. 02-217-735
Add to Cart
Add to Cart
For use with Fisherbrand™ Incubating Rockers, Open Air Rockers and Incubating Wavers.
  • Dimpled mat designed to hold tubes and vials of different sizes
  • Stacking tray (Cat. No. 02-217-736) attaches to Open Air Rocker unit to add second tier, mounts 3.5 in. (8.9cm) above lower tray, includes rubber mat and all hardware
TRUSTED_SUSTAINABILITY
Specifications
Product Type Platform Mat
Material Rubber
For Use With (Equipment) Rockers