Learn More
Grainger JAMCO General Purpose Platform Truck, Steel Deck, 1,200 lb Capacity, 72 in x 30 in, 1 ea
Catalog No. 502788199
Encompass_Preferred
This general purpose platform truck features a solid 12 ga steel deck designed for efficient material handling of heavy or bulky items. Its low-profile 12 in. deck height facilitates easier loading and unloading, while the robust steel construction with a durable powder-coated finish ensures long-term reliability in demanding environments.
  • Load capacity of 1,200 lb
  • Deck dimensions: 72 in. L x 30 in. W
  • Equipped with four 8 in. semipneumatic rubber caster wheels (2 rigid, 2 swivel) for smooth transport
  • Includes a removable raised handle with a single-end pocket
  • Solid, smooth surface with a flush lip design
  • Ships unassembled
Catalog No. 50-278-8199 Supplier Grainger Supplier No. 16C634
This general purpose platform truck features a solid 12 ga steel deck designed for efficient material handling of heavy or bulky items. Its low-profile 12 in. deck height facilitates easier loading and unloading, while the robust steel construction with a durable powder-coated finish ensures long-term reliability in demanding environments.
  • Load capacity of 1,200 lb
  • Deck dimensions: 72 in. L x 30 in. W
  • Equipped with four 8 in. semipneumatic rubber caster wheels (2 rigid, 2 swivel) for smooth transport
  • Includes a removable raised handle with a single-end pocket
  • Solid, smooth surface with a flush lip design
  • Ships unassembled