Learn More
Grainger Jamco Platform Truck, Steel, 2,000 lb. Capacity, 48 in. x 24 in., 1 ea.
Catalog No. 502789623
Encompass_Preferred
This heavy-duty platform truck is engineered for industrial transport, featuring a robust 12 gauge steel deck and an all-welded frame construction for maximum durability. Designed for a 2,000 lb. evenly distributed load capacity, it offers reliable performance in demanding environments. The truck includes a removable handle and a flush deck design to facilitate easy loading and unloading of goods.
  • Durable 12 gauge steel deck and caster mounts
  • Flush deck design with 1.5 in. lips for easy loading
  • Removable tubular handle with smooth radius bend
  • Four 5 in. phenolic casters in a configuration of two swivel and two rigid for superior tracking
  • Powder coated finish for increased resistance to wear
  • All-welded construction excluding casters and handle
Catalog No. 50-278-9623 Supplier Grainger Supplier No. 16C688
This heavy-duty platform truck is engineered for industrial transport, featuring a robust 12 gauge steel deck and an all-welded frame construction for maximum durability. Designed for a 2,000 lb. evenly distributed load capacity, it offers reliable performance in demanding environments. The truck includes a removable handle and a flush deck design to facilitate easy loading and unloading of goods.
  • Durable 12 gauge steel deck and caster mounts
  • Flush deck design with 1.5 in. lips for easy loading
  • Removable tubular handle with smooth radius bend
  • Four 5 in. phenolic casters in a configuration of two swivel and two rigid for superior tracking
  • Powder coated finish for increased resistance to wear
  • All-welded construction excluding casters and handle