Learn More
Grainger JAMCO Platform Truck, Steel, 2,000 lb. Capacity, 1 ea.
Catalog No. 502791336
Encompass_Preferred
This standard platform truck is designed for heavy-duty material transport, offering a robust solution for moving larger items in industrial environments. The truck features a solid steel deck with a powder-coated finish for durability and is engineered to support loads up to 2,000 lbs.
  • Constructed with a 12-gauge steel deck for maximum strength
  • Equipped with four removable corner post handles to facilitate the transport of longer materials
  • Includes four 6 in. phenolic casters in a configuration of two rigid and two swivel for easy maneuverability
  • Powder-coated finish provides a durable, smooth surface
  • Comes unassembled for flexible on-site setup
Catalog No. 50-279-1336 Supplier Grainger Supplier No. 16C589
This standard platform truck is designed for heavy-duty material transport, offering a robust solution for moving larger items in industrial environments. The truck features a solid steel deck with a powder-coated finish for durability and is engineered to support loads up to 2,000 lbs.
  • Constructed with a 12-gauge steel deck for maximum strength
  • Equipped with four removable corner post handles to facilitate the transport of longer materials
  • Includes four 6 in. phenolic casters in a configuration of two rigid and two swivel for easy maneuverability
  • Powder-coated finish provides a durable, smooth surface
  • Comes unassembled for flexible on-site setup