Learn More
Grainger JAMCO Platform Truck, Steel, 2,000 lb Capacity, 72 in Length, 1 ea
Catalog No. 502789897
Encompass_Preferred
This heavy-duty platform truck is engineered for the transport of long or oversized materials such as pipe, bar stock, and rolls. Its open-sided design, combined with removable corner posts, allows for versatile loading and unloading while providing essential containment during transport. The truck features a low-profile deck to minimize lift distances for bulky items.
  • Load capacity of 2,000 lb
  • 12 ga steel deck with powder coated finish
  • Overall dimensions of 77 in L x 31 in W x 40 in H
  • Equipped with four removable corner posts for steering and load retention
  • Four 6 in phenolic casters in a configuration of two rigid and two swivel
  • Unassembled delivery
Catalog No. 50-278-9897 Supplier Grainger Supplier No. 16C590
This heavy-duty platform truck is engineered for the transport of long or oversized materials such as pipe, bar stock, and rolls. Its open-sided design, combined with removable corner posts, allows for versatile loading and unloading while providing essential containment during transport. The truck features a low-profile deck to minimize lift distances for bulky items.
  • Load capacity of 2,000 lb
  • 12 ga steel deck with powder coated finish
  • Overall dimensions of 77 in L x 31 in W x 40 in H
  • Equipped with four removable corner posts for steering and load retention
  • Four 6 in phenolic casters in a configuration of two rigid and two swivel
  • Unassembled delivery